Learn More
Grace Davison™ HPLC Column Visionht™
Brand:  Grace Davison™ 5148864
Product Code. 12121290
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Maximise speed and resolution
  • High throughput technologies for all systems
  • Simplify separation strategies
  • Complementary phase chemistries
  • Improve laboratory efficiency
  • Seamless method transfer between UHPLC, HPLC and Prep
  • UHPLC columns rated to 16,000psi for compatibility on all systems
  • Low dead volume column hardware used for all UHPLC columns
  • UltraMD UHPLC method development kits available containing C18 classic, C18 polar, C18 high load and C18 basic columns in selected dimension
  • Other sizes available