missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ HPLC Air Valve, for PFAS Analytic
GREENER_CHOICE

Product Code. 18686458 Shop All SCAT Europe Products
Change view
Click to view available options
Quantity:
1 Ea.
10 Pk.
8 Pk.
Unit Size:
1 sheet
10 sets
8 sets
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18686458 1 Ea. 1 sheet
18696488 8 Pk. 8 sets
18606498 10 Pk. 10 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18686458 Supplier SCAT™ Supplier No. 317011

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The SCAT air valve blocks harmful vapours and ensures safe pressure equalisation in the storage tanks of your HPLC. Solvent mixtures remain stable: thanks to closed system without evaporation.

  • With Luer-Lock connection for gassing the solvents and change indicator for timely change.
  • Made of PP for PFAS analysis of polyfluorinated chemicals (formerly: PFC)
  • UNF 1/4 in. 28G thread size fits to any SCA PFAS-Analytic Safety Cap for PFAS Analysis

Specifications

For Use With (Equipment) SCAT Safety Caps for PFAS-Analytic
Material PP
Thread Size UNF 1/4 in. 28G
For Use With (Application) HPLC analysis
Type Valve
Quantity 1 Ea.
Color Red, Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.