missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HPi2 Monoclonal antibody specifically detects HPi2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | HPi2 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | HIC1-2B4.2B |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | human pancreatic endocrine islet cell |
| Host Species | Mouse |
| Immunogen | Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells. |
| Purification Method | Tissue culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?