missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPGD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11035-1-AP-150UL
This item is not returnable.
View return policy
Description
Prostaglandin inactivation. Contributes to the regulation of events that are under the control of prostaglandin levels. Catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. Inhibits in vivo proliferation of colon cancer cells.Specifications
| HPGD | |
| Polyclonal | |
| Unconjugated | |
| HPGD | |
| 15 PGDH, HPGD, PGDH, PGDH1, Prostaglandin dehydrogenase 1, SDR36C1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3248 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P15428 | |
| HPGD | |
| HPGD Fusion Protein Ag1499 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction