missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HPDL Recombinant Rabbit Monoclonal Antibody (K01_1S49)

Product Code. 30828853
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828853 50 μL
30825575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828853 Supplier Invitrogen™ Supplier No. MA565269
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

HPDL Recombinant Monoclonal Antibody for Western Blot

May have dioxygenase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPDL
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1S49
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene HPDL
Gene Accession No. Q96IR7
Gene Alias 4-HPPD-L; 4-hydroxyphenylpyruvate dioxygenase like; 4-hydroxyphenylpyruvate dioxygenase-like protein; GLOXD1; glyoxalase domain containing 1; glyoxalase domain-containing protein 1; HPDL; RP4-534D1.1
Gene Symbols HPDL
Host Species Rabbit
Immunogen A synthetic peptide of human HPDL.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84842
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.