Learn More
HPDL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556898
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SPGEDPELGL EMTAGFGLGG LRLTALQAQP GSIVPTLVLA ESLPGATTRQ DQVEQFLARH KGPGLQHVGL YTPNIVEATE GVATAGGQFL AP Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 75%.
May have dioxygenase activity.Specifications
HPDL | |
Polyclonal | |
Unconjugated | |
HPDL | |
4-HPPD-L; 4-hydroxyphenylpyruvate dioxygenase like; 4-hydroxyphenylpyruvate dioxygenase-like protein; GLOXD1; glyoxalase domain containing 1; glyoxalase domain-containing protein 1; HPDL; RP4-534D1.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84842 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96IR7 | |
HPDL | |
Recombinant protein corresponding to Human HPDL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only