missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HPD Monoclonal Antibody (D1), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA550968

 View more versions of this product

Product Code. 17910353

  • £161.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The inhibitor of apoptosis (IAP) family of proteins regulates programmed cell death triggered by various stimuli. All IAPs have at least one baculovirus IAP repeat (BIR) motif that is essential for their anti-apoptotic activity (1-3). Recently, a serine protease has been isolated, which is released from the mitochondria upon induction of apoptosis by apoptotic stimuli. This protein called Omi/HtrA2 is a mitochondrial protein that binds to IAP (4). Like Smac/DIABLO, the mature Omi protein contains a conserved IAP-binding motif (AVPS) at its N terminus. The deregulated expression of Omi in the cytoplasm of mammalian cells induces apoptosis indicating that Omi could participate in the mitochondrial apoptotic pathway.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HPD
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
P32754, Q02110
Hpd
Recombinant Human HPD protein, Thr2-Met393 (Accession #P32754).
20 μL
Primary
Human, Pig
Antibody
IgG1 κ
Immunohistochemistry (Paraffin), Western Blot
D1
Unconjugated
Hpd
4H GLOD3; 4-H HPPDASE; 4HPPD; 4-HPPD; 4-hydroxyphenylpyruvate dioxygenase; 4-hydroxyphenylpyruvic acid dioxygenase; 4-hydroxyphenylpyruvic acid oxidase; f Alloantigen; F liver antigen; f protein; Fla; Flp; GLOD3; glyoxalase domain containing 3; HPD; Hppd; HPPDASE; Laf; PPD
Mouse
Protein A/G
RUO
3242, 397443
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.