missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPCL Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18417R
This item is not returnable.
View return policy
Description
HPCL Polyclonal antibody specifically detects HPCL in Human, Mouse samples. It is validated for Western Blot
Specifications
| HPCL | |
| Polyclonal | |
| Western Blot | |
| 1600020H07Rik, 2,2-HPCL, 2-hydroxyacyl-CoA lyase 1, 2-hydroxyphytanoyl-CoA lyase2-hydroxyphytanol-CoA lyase, EC 4.1, HPCL, PHYH2FLJ55041, Phytanoyl-CoA 2-hydroxylase 2, phytanoyl-CoA hydroxylase 2 | |
| Recognizes region between residues Between Between 528 and 578. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 26061 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction