missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HPa2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560773
This item is not returnable.
View return policy
Description
Immunogen sequence: GKRTDFLQFQ NLRNPAKSRG GPGPDYYLKN YEDDIVRSDV ALDKQKGCKI AQHPDVMLEL QREKAAQMHL VLLKQQFSNT Y Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 25%.
This gene encodes a heparanase enzyme. The encoded protein is a endoglycosidase that degrades heparin sulfate proteoglycans located on the extracellular matrix and cell surface. This protein may be involved in biological processes involving remodeling of the extracellular matrix including angiogenesis and tumor progression. Alternate splicing results in multiple transcript variants.
Specifications
| HPa2 | |
| Polyclonal | |
| Unconjugated | |
| HPSE2 | |
| Gm968; heparanase 2; heparanase 2 (inactive); heparanase 3; heparanase-2; heparanase-like protein; Hpa2; HPR2; Hpse2; Inactive heparanase-2; UFS; UFS1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 60495 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WWQ2 | |
| HPSE2 | |
| Recombinant protein corresponding to Human HPa2. Recombinant protein control fragment (Product #RP-97170). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction