missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPa2 Antibody (DHIC2-2B4) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-18950JF585
This item is not returnable.
View return policy
Description
HPa2 Monoclonal antibody specifically detects HPa2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
Specifications
| HPa2 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| human pancreatic alpha cell | |
| Human pancreatic enriched alpha cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| DHIC2-2B4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction