missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HPa2 Monoclonal antibody specifically detects HPa2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | HPa2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | DHIC2-2B4 |
| Conjugate | Janelia Fluor 549 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | human pancreatic alpha cell |
| Host Species | Mouse |
| Immunogen | Human pancreatic enriched alpha cells |
| Purification Method | Tissue culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?