Learn More
Invitrogen™ HPa1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117025
Description
Positive Samples: Jurkat, DU145, 293T Immunogen sequence: QDVVDLDFFT QEPLHLVSPS FLSVTIDANL ATDPRFLILL GSPKLRTLAR GLSPAYLRFG GTKTDFLIFD PKKESTFEER SYWQSQVNQD ICKYGSIPPD VEEKLRLEWP YQEQLLLREH YQKKFKNSTY SRSSVDVLYT FANCSGLDLI FGLNALLRTA DLQWNSSNAQ LLLDYCSSKG YNISWELGNE PNSFLKKADI FINGSQLGED FIQLHKLLRK STFKNAKLYG PDVGQPRRKT.
Stem cells offer a therapeutic promise for many diseases. In the case of diabetes, stem cells offer the promise that beta cells may be generated ex vivo for the possible transplantation and cure of Type I diabetes (Couri CE, et al. ). For this to happen, stem cells will need to be isolated and expanded, they will need to be differentiated toward the beta cell lineage, and the differentiated cell progeny used for transplantation may need to be isolated from complex cell mixtures following differentiation. Appropriate markers targeting cell surface molecules on developing and mature pancreatic cells will be required for such research.
Specifications
| HPa1 | |
| Polyclonal | |
| Unconjugated | |
| HPSE | |
| Endo-glucoronidase; endoglycosidase heparanase; HEP; Heparanase; Heparanase 50 kDa subunit; Heparanase 8 kDa subunit; Heparanase-1; heparanase-like; Hpa; HPA1; HPR1; HPSE; HPSE1; HSE1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10855, 15442, 64537 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.93 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6YGZ1, Q71RP1, Q9Y251 | |
| HPSE | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.