missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HP1 gamma Recombinant Rabbit Monoclonal Antibody (K01_1S48)
Description
Immunogen sequence: FTDADNTWEP EENL.
At the nuclear envelope, the nuclear lamina and heterochromatin are adjacent to the inner nuclear membrane. The protein encoded by this gene binds DNA and is a component of heterochromatin. This protein also can bind lamin B receptor, an integral membrane protein found in the inner nuclear membrane. The dual binding functions of the encoded protein may explain the association of heterochromatin with the inner nuclear membrane.
Specifications
Specifications
| Antigen | HP1 gamma |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1S48 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | Cbx3 |
| Gene Accession No. | Q13185 |
| Gene Alias | CBX3; chromobox 3; chromobox homolog 3; chromobox homolog 3 (HP1 gamma homolog, Drosophila); chromobox protein homolog 3; chromobox-like protein 3; HECH; heterochromatin protein 1 gamma; heterochromatin protein 1 homolog gamma; heterochromatin protein HP1 gamma; heterochromatin-like protein 1; HGNC:1553; HP1 gamma; HP1 gamma homolog; HP1g; HP1-GAMMA; HP1Hs-gamma; HPy; M32; Modifier 2 protein; OTTHUMP00000202463 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?