missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HP1 gamma Recombinant Rabbit Monoclonal Antibody (K01_1S47)

Product Code. 30826355
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826355 100 μL
30826721 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826355 Supplier Invitrogen™ Supplier No. MA563615

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

HP1 gamma Recombinant Monoclonal Antibody for IHC (P), Western Blot

At the nuclear envelope, the nuclear lamina and heterochromatin are adjacent to the inner nuclear membrane. The protein encoded by this gene binds DNA and is a component of heterochromatin. This protein also can bind lamin B receptor, an integral membrane protein found in the inner nuclear membrane. The dual binding functions of the encoded protein may explain the association of heterochromatin with the inner nuclear membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HP1 gamma
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1S47
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene Cbx3
Gene Accession No. P23198, Q13185
Gene Alias CBX3; chromobox 3; chromobox homolog 3; chromobox homolog 3 (HP1 gamma homolog, Drosophila); chromobox protein homolog 3; chromobox-like protein 3; HECH; heterochromatin protein 1 gamma; heterochromatin protein 1 homolog gamma; heterochromatin protein HP1 gamma; heterochromatin-like protein 1; HGNC:1553; HP1 gamma; HP1 gamma homolog; HP1g; HP1-GAMMA; HP1Hs-gamma; HPy; M32; Modifier 2 protein; OTTHUMP00000202463
Gene Symbols Cbx3
Host Species Rabbit
Immunogen A synthetic peptide of mouse HP1 gamma.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11335, 12417, 297093
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.