missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HP1 alpha Recombinant Human Monoclonal Antibody (RAB-C133)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551856
This item is not returnable.
View return policy
Description
This antibody recognizes CBX5 (Chromobox protein homolog 5). It binds to a folded domain, amino acids 18-75. CBX5 is a component of heterochromatin that recognizes and binds histone H3 tails methylated at 'Lys-9' (H3K9me), leading to epigenetic repression.
Heterochromatin protein-1 (HP1) is a methyl-lysine binding protein localized at heterochromatin sites, where it mediates gene silencing. It has been shown that mammalian methyltransferases that selectively methylate histone H3 on lysine-9 generate a binding site for HP1 proteins, a family of heterochromatic adaptor molecules implicated in both gene silencing and supranucleosomal chromatin structure. High-affinity in vitro recognition of a methylated histone H3 peptide by HP1 requires a functional chromodomain. Thus, the HP1 chromodomain is a specific interaction motif for the methyl epitope on lysine-9 of histone H3. In vivo, heterochromatin association of HP1 proteins is lost in Suv39h double-null primary mouse fibroblasts but is restored after reintroduction of a catalytically active SUV39H1 HMTase.
Specifications
| HP1 alpha | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P45973 | |
| Cbx5 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing CBX5 protein under non-denaturing conditions. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| F(ab) κ |
| ChIP Assay, ELISA, Mass Spectrometry | |
| RAB-C133 | |
| Unconjugated | |
| Cbx5 | |
| 2610029O15Rik; antigen p25; C75991; Cbx5; Cbx5 antibody; chromobox 5; chromobox homolog 5; chromobox homolog 5 (HP1 alpha homolog, Drosophila); chromobox protein homolog 5; chromobox-like protein 5; epididymis luminal protein 25; HEL25; heterochromatin protein 1 alpha; heterochromatin protein 1 homolog alpha; heterochromatin protein 1-alpha; HGNC:1555; HP1; HP1 alpha; HP-1 alpha antibody; HP1 alpha homolog; Hp1a; Hp1alpha; HP1-ALPHA; HP1Hs alpha; HP1Hs-alpha; mHP1(alpha) | |
| Human | |
| Metal-chelate chromatography | |
| RUO | |
| 23468 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction