missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HP1 alpha Antibody (RAB-C133) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20127-0.2mg
This item is not returnable.
View return policy
Description
HP1 alpha Monoclonal antibody specifically detects HP1 alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)
Specifications
| HP1 alpha | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation | |
| RAB-C133 | |
| Unconjugated | |
| Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing CBX5 protein under non-denaturing conditions. | |
| 0.2 mg | |
| Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 23468 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction