missing translation for 'onlineSavingsMsg'
Learn More

HOXD9 Antibody (OTI1H3), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

£195.00 - £417.00

Specifications

Antigen HOXD9
Clone OTI1H3
Dilution Western Blot 1:2000, Immunohistochemistry 1:150
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306311
View Documents
Novus Biologicals
NBP3-45002-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306676
View Documents
Novus Biologicals
NBP3-45002-30ul
30 μL
£195.00
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HOXD9 Monoclonal antibody specifically detects HOXD9 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications

Specifications

HOXD9
Western Blot 1:2000, Immunohistochemistry 1:150
Monoclonal
Purified
RUO
PBS (pH 7.3) 1% BSA, 50% glycerol
3235
IgG1
>95%
Antibody
OTI1H3
Immunohistochemistry, Western Blot
Unconjugated
Mouse
Human
homeo box D9, homeobox D9, Homeobox protein Hox-4C, Homeobox protein Hox-5.2, homeobox protein Hox-D9, HOX4, Hox-4.3, mouse, homolog of, HOX4CHox-4.3, Hox-5.2
Human recombinant protein fragment corresponding to amino acids 109-352 of human HOXD9 (NP_055028) produced in E.coli.
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.