missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXD8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120500
This item is not returnable.
View return policy
Description
Positive test controls include: SKOV3, Mouse ovary, Rat ovary. Immunogen sequence: RQTYSRFQTL ELEKEFLFNP YLTRKRRIEV SHALALTERQ VKIWFQNRRM KWKKENNKDK FPVSRQEVKD GETKKEAQEL EEDRAEGLTN.
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located in a cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. In addition to effects during embryogenesis, this particular gene may also play a role in adult urogenital tract function.
Specifications
| HOXD8 | |
| Polyclonal | |
| Unconjugated | |
| HOXD8 | |
| 4921540P06Rik; AI047735; homeo box 4E; homeo box D8; homeobox D8; homeobox protein 5.4; homeobox protein Hox-4.3; Homeobox protein Hox-4E; homeobox protein Hox-5.4; Homeobox protein Hox-D8; homeobox protein Hox-D8-like; HOX4; Hox-4.3; Hox-4.5; HOX4E; HOX5.4; Hox-5.4; HOXD8; Hoxd-8; LOC100912936 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100912936, 15437, 3234 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.91 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P13378, P23463 | |
| HOXD8 | |
| A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human HOXD8 (NP_001186675.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction