missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565867
This item is not returnable.
View return policy
Description
Immunogen sequence: PCEEYLQGGYL GEQGADYYGG GAQGADFQPP GLYPRPDFGE QPFGGSG Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
Specifications
| HOXD4 | |
| Polyclonal | |
| Unconjugated | |
| HOXD4 | |
| 6030436D05Rik; HHO.C13; homeo box D4; homeobox D4; homeobox protein HHO.C13; Homeobox protein Hox-4.2; homeobox protein Hox-4B; Homeobox protein Hox-5.1; homeobox protein Hox-D4; HOX4; Hox-4.2; Hox-4.2, mouse, homolog of homeo box X; HOX4B; Hox-5.1; HOXD4; Hoxd-4; LOW QUALITY PROTEIN: homeobox protein Hox-D4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3233 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P09016 | |
| HOXD4 | |
| Recombinant Human HOXD4. Recombinant protein control fragment (Product #RP-106538). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction