missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXD4 Monoclonal Antibody (1H7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524409
This item is not returnable.
View return policy
Description
Peptide Sequence: MVMSSYMVNS KYVDPKFPPC EEYLQGGYLG EQGADYYGGG AQGADFQPPG LYPRPDFGEQ PF.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
Specifications
| HOXD4 | |
| Monoclonal | |
| Unconjugated | |
| HOXD4 | |
| 6030436D05Rik; HHO.C13; homeo box D4; homeobox D4; homeobox protein HHO.C13; Homeobox protein Hox-4.2; homeobox protein Hox-4B; Homeobox protein Hox-5.1; homeobox protein Hox-D4; HOX4; Hox-4.2; Hox-4.2, mouse, homolog of homeo box X; HOX4B; Hox-5.1; HOXD4; Hoxd-4; LOW QUALITY PROTEIN: homeobox protein Hox-D4 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3233 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1H7 | |
| PBS with no preservative; pH 7.4 | |
| P09016 | |
| HOXD4 | |
| HOXD4 (NP_055436.2, 1 a.a. ~ 62 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction