missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22684-1-AP-20UL
This item is not returnable.
View return policy
Description
The Hox proteins are a family of transcription factors that play a role in development and cellular differentiation by regulating downstream target genes. Specifically, the Hox proteins direct DNA-protein and protein-protein interactions that assist in determining the morphologic features associated with the anterior-posterior body axis. Hox proteins are involved in controlling axial patterning, leukemias and hereditary malformations. HoxD12 (homeobox D12), also known as HOX4H, is a 275 amino acid protein that localizes to the nucleus and contains one homeobox DNA-binding domain. One of several members of the homeobox superfamily, HoxD12 functions as a sequencespecific transcription factor that is important for the correct positioning of developing limb buds on the anterior-posterior axis.Specifications
| HOXD12 | |
| Polyclonal | |
| Unconjugated | |
| HOXD12 | |
| homeobox D12, Homeobox protein Hox 4H, Homeobox protein Hox D12, HOX4H, HOXD12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15432, 3238 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P23812, P35452 | |
| Hoxd12 | |
| HOXD12 Fusion Protein Ag18577 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction