missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXD12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599289
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HOXD12.
The Hox proteins are a family of transcription factors that play a role in development and cellular differentiation by regulating downstream target genes. Specifically, the Hox proteins direct DNA-protein and protein-protein interactions that assist in determining the morphologic features associated with the anterior-posterior body axis. Hox proteins are involved in controlling axial patterning, leukemias and hereditary malformations. HoxD12 (homeobox D12), also known as HOX4H, is a 275 amino acid protein that localizes to the nucleus and contains one homeobox DNA-binding domain. One of several members of the homeobox superfamily, HoxD12 functions as a sequencespecific transcription factor that is important for the correct positioning of developing limb buds on the anterior-posterior axis.
Specifications
| HOXD12 | |
| Polyclonal | |
| Unconjugated | |
| HOXD12 | |
| homeo box D12; homeobox D12; homeobox protein Hox-4.7; Homeobox protein Hox-4H; homeobox protein Hox-5.6; Homeobox protein Hox-D12; Hox 4H; Hox-4.7; Hox-4.7, mouse, homolog of; HOX4H; Hox-5.6; Hoxd12; Hoxd-12 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15432, 3238 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P23812, P35452 | |
| HOXD12 | |
| A synthesized peptide derived from human HOXD12(Accession P35452), corresponding to amino acid residues A68-S118. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction