missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXD10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116181
This item is not returnable.
View return policy
Description
HOXD10 Polyclonal Antibody for Western Blot
This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox D genes located on chromosome 2. The encoded nuclear protein functions as a sequence-specific transcription factor that is expressed in the developing limb buds and is involved in differentiation and limb development. Mutations in this gene have been associated with Wilm's tumor and congenital vertical talus and/or a foot deformity resembling that seen in Charcot-Marie-Tooth disease.
Specifications
| HOXD10 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P28358, P28359, Q90469 | |
| HOXD10, hoxd10a | |
| KLH conjugated synthetic peptide between 305-334 amino acids from the C-terminal region of human HOXD10. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Zebrafish | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| HOXD10 | |
| AI385591; AI874987; d10; homeo box 4D; homeo box D10; homeo box D10a; homeobox D10; homeobox D10a; homeobox gene D-10; homeobox protein Hox-4.5; Homeobox protein Hox-4D; homeobox protein Hox-4E; Homeobox protein Hox-5.3; homeobox protein Hox-C10; Homeobox protein Hox-D10; homeobox protein Hox-D10a; homeobox protein Hox-D11; homeobox protein Hox-D10; HOX4; Hox-4.4; Hox-4.5; Hox-45; HOX4D; HOX4E; Hox-5.3; hoxc10; hox-C10; Hoxc10 protein; Hoxd10; HOX-D10; hoxd-10; hoxd10a; hoxd10a protein; z-82 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 15430, 30404, 3236 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction