missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD10 Antibody (OTI1D11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70920CL1
This item is not returnable.
View return policy
Description
HOXD10 Monoclonal antibody specifically detects HOXD10 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| HOXD10 | |
| Monoclonal | |
| CoraFluor 1 | |
| homeo box 4D, homeo box D10, homeobox D10, Homeobox protein Hox-4D, Homeobox protein Hox-4E, homeobox protein Hox-D10, Hox-4.4, HOX4DHOX4, HOX4E | |
| Human recombinant protein fragment corresponding to amino acids 80-340 of human HOXD10 (NP_002139) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI1D11 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 3236 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction