missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXC9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567618
This item is not returnable.
View return policy
Description
HOXC9 Polyclonal Antibody for IHC (P), Western Blot
HOXC9 belongs to the homeobox family. The homeobox family is a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms.This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12.
Specifications
| HOXC9 | |
| Polyclonal | |
| Unconjugated | |
| HOXC9 | |
| homeo box C9; homeobox C9; homeobox protein Hox-3.2; Homeobox protein Hox-3B; Homeobox protein Hox-C9; HOX3; Hox-3.2; HOX3B; Hoxc8; Hoxc9; HOXC-9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15427, 3225 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P09633, P31274 | |
| HOXC9 | |
| Synthesized peptide derived from human HOXC9. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction