Learn More
Hoxc5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555421
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PGHAPGRDEA APLNPGMYSQ KAARPALEER AKSSGEIKEE QAQTGQPAGL SQPPAPPQIY PWMT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.Specifications
Hoxc5 | |
Polyclonal | |
Unconjugated | |
HOXC5 | |
AI181837; CP11; homeo box C5; homeobox C5; homeobox protein CP11; Homeobox protein Hox-3.4; homeobox protein Hox-3D; Homeobox protein Hox-6.2; homeobox protein Hox-C5; HOX3; Hox-3.4; Hox-34; HOX3D; Hox-6.2; HOXC5; Hoxc-5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3222 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q00444 | |
HOXC5 | |
Recombinant protein corresponding to Human Hoxc5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only