missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14321-1-AP-150UL
This item is not returnable.
View return policy
Description
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.Specifications
| HOXC4 | |
| Polyclonal | |
| Unconjugated | |
| HOXC4 | |
| cp19, homeobox C4, Homeobox protein CP19, Homeobox protein Hox 3E, Homeobox protein Hox C4, HOX3, HOX3E, HOXC4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15423, 24459, 3221 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09017, P18865, Q08624 | |
| HOXC4 | |
| HOXC4 Fusion Protein Ag5563 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction