missing translation for 'onlineSavingsMsg'
Learn More

HOXC4 Antibody (OTI6B11) [PE], Novus Biologicals™

Product Code. 30688789 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688789 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688789 Supplier Novus Biologicals Supplier No. NBP345008PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HOXC4 Monoclonal antibody specifically detects HOXC4 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC4
Applications Western Blot
Classification Monoclonal
Clone OTI6B11
Conjugate PE
Formulation PBS
Gene Alias homeo box 3E, homeo box C4, homeobox C4, Homeobox protein CP19, Homeobox protein Hox-3E, homeobox protein Hox-C4, HOX3, HOX3Ecp19
Host Species Mouse
Immunogen Full length human recombinant protein of human HOXC4 (NP_705897) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3221
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.