missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC4 Antibody (OTI3F3), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45017-30ul
This item is not returnable.
View return policy
Description
HOXC4 Monoclonal antibody specifically detects HOXC4 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| HOXC4 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| 3221 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
| Western Blot | |
| OTI3F3 | |
| Western Blot 1:2000 | |
| homeo box 3E, homeo box C4, homeobox C4, Homeobox protein CP19, Homeobox protein Hox-3E, homeobox protein Hox-C4, HOX3, HOX3Ecp19 | |
| Full length human recombinant protein of human HOXC4(NP_705897) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction