missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC4 Antibody (OTI1F6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£195.00 - £417.00
Specifications
| Antigen | HOXC4 |
|---|---|
| Clone | OTI1F6 |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:2000 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306722
|
Novus Biologicals
NBP3-45006-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306569
|
Novus Biologicals
NBP3-45006-30ul |
30 μL |
£195.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXC4 Monoclonal antibody specifically detects HOXC4 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| HOXC4 | |
| Western Blot 1:2000, Immunohistochemistry 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 3221 | |
| IgG2a | |
| >95% | |
| Antibody |
| OTI1F6 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| homeo box 3E, homeo box C4, homeobox C4, Homeobox protein CP19, Homeobox protein Hox-3E, homeobox protein Hox-C4, HOX3, HOX3Ecp19 | |
| Full length human recombinant protein of human HOXC4 (NP_705897) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title