missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£222.00 - £484.00
Specifications
| Antigen | HOXC4 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18217614
|
Novus Biologicals
NBP2-56195 |
100 μL |
£484.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611568
|
Novus Biologicals
NBP2-56195-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXC4 Polyclonal specifically detects HOXC4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| HOXC4 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| homeo box 3E, homeo box C4, homeobox C4, Homeobox protein CP19, Homeobox protein Hox-3E, homeobox protein Hox-C4, HOX3, HOX3Ecp19 | |
| HOXC4 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 3221 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PNTKVRSAPPAGAAPSTLSAATPGTSEDHSQSATPPEQQRAEDITRL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title