missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HOXC4 Monoclonal antibody specifically detects HOXC4 in Human samples. It is validated for ELISA,Western Blot,KnockDown
Specifications
Specifications
| Antigen | HOXC4 |
| Applications | ELISA, Western Blot, KnockDown |
| Classification | Monoclonal |
| Clone | 1E9 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Knockdown Validated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_705897 |
| Gene Alias | homeo box 3E, homeo box C4, homeobox C4, Homeobox protein CP19, Homeobox protein Hox-3E, homeobox protein Hox-C4, HOX3, HOX3Ecp19 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?