missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC13 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£163.00 - £380.00
Specifications
| Antigen | HOXC13 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18352244
|
Novus Biologicals
NBP3-04679-20UL |
20 μL |
£163.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18363127
|
Novus Biologicals
NBP3-04679-100UL |
100 μL |
£380.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXC13 Polyclonal specifically detects HOXC13 in Human samples. It is validated for Western Blot.Specifications
| HOXC13 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 3229 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| homeo box 3G, homeo box C13, homeobox C13, Homeobox protein Hox-3G, homeobox protein Hox-C13, HOX3, HOX3GNUP98/HOXC13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-330 of human HOXC13 (NP_059106.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title