missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXC12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103893
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HOXC12.
The Homeobox (Hox) proteins are a family of transcription factors that play a role in development and cellular differentiation by regulating downstream target genes. Specifically, the Hox proteins direct DNA-protein and protein-protein interactions that assist in determining the morphologic features associated with the anterior-posterior body axis. Hox proteins are involved in controlling axial patterning, leukemias and hereditary malformations. In mammals, there are four gene clusters encoding Hox proteins, namely HoxA, HoxB, HoxC and HoxD, that are located on different chromosomes, each consisting of 9-11 tandemly arranged genes. HoxC12 (Homeobox C12), also known as Hox3, Hox3F or HOC3F, is a member of the Abd-B homeobox (Hox) family and is encoded by a gene from the HoxC cluster. HoxC12 is a 282 amino acid long nuclear protein that contains one homeobox DNA-binding domain.
Specifications
| HOXC12 | |
| Polyclonal | |
| Unconjugated | |
| HOXC12 | |
| HOC3F; homeo box C12; homeobox C12; homeobox protein Hox-3.8; Homeobox protein Hox-3F; Homeobox protein Hox-C12; Homeobox protein Hox-C12 (Hox-3F); HOX3; Hox-3.8; HOX3F; HOXC12; Hoxc-12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15421, 300262, 3228 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P31275, Q8K5B8 | |
| HOXC12 | |
| A synthesized peptide derived from human HOXC12(Accession P31275), corresponding to amino acid residues P65-D115. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction