missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXC12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563152
This item is not returnable.
View return policy
Description
Immunogen sequence: LLNPGFVGPL VNIHTGDTFY FPNFRASGAQ LPGLPSLSYP RRDNVCSLSW PSAEPCNGYP QPYLGSPVSL NPPF Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
The Homeobox (Hox) proteins are a family of transcription factors that play a role in development and cellular differentiation by regulating downstream target genes. Specifically, the Hox proteins direct DNA-protein and protein-protein interactions that assist in determining the morphologic features associated with the anterior-posterior body axis. Hox proteins are involved in controlling axial patterning, leukemias and hereditary malformations. In mammals, there are four gene clusters encoding Hox proteins, namely HoxA, HoxB, HoxC and HoxD, that are located on different chromosomes, each consisting of 9-11 tandemly arranged genes. HoxC12 (Homeobox C12), also known as Hox3, Hox3F or HOC3F, is a member of the Abd-B homeobox (Hox) family and is encoded by a gene from the HoxC cluster. HoxC12 is a 282 amino acid long nuclear protein that contains one homeobox DNA-binding domain.
Specifications
| HOXC12 | |
| Polyclonal | |
| Unconjugated | |
| HOXC12 | |
| HOC3F; homeo box C12; homeobox C12; homeobox protein Hox-3.8; Homeobox protein Hox-3F; Homeobox protein Hox-C12; Homeobox protein Hox-C12 (Hox-3F); HOX3; Hox-3.8; HOX3F; HOXC12; Hoxc-12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3228 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P31275 | |
| HOXC12 | |
| Recombinant protein corresponding to Human HOXC12. Recombinant protein control fragment (Product #RP-97593). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction