Learn More
Invitrogen™ HOXC11 Monoclonal Antibody (OTI3E10)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525460
Product Code. 15935064
  • £474.00 / 100µL
Description
HOXC11 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene binds to a promoter element of the lactase-phlorizin hydrolase. It also may play a role in early intestinal development. An alternatively spliced variant encoding a shorter isoform has been described but its full-length nature has not been determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HOXC11
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O43248
HOXC11
Human recombinant protein fragment corresponding to amino acids 1-304 of HOXC11 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI3E10
Unconjugated
HOXC11
homeo box C11; homeobox C11; homeobox protein Hox-3.7; Homeobox protein Hox-3H; homeobox protein Hox-C11; homeobox protein Hox-C11-like; Hox-3.7; HOX3H; HOXC11; Hoxc-11; LOC100911859; LOW QUALITY PROTEIN: homeobox protein Hox-C11; MGC4906
Mouse
Affinity Chromatography
RUO
3227
-20°C, Avoid Freeze/Thaw Cycles