missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXC10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531078
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, HepG2, mouse muscle. Predicted reactivity: Mouse (97%), Rat (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The protein level is controlled during cell differentiation and proliferation, which may indicate this protein has a role in origin activation.
Specifications
| HOXC10 | |
| Polyclonal | |
| Unconjugated | |
| HOXC10 | |
| AI506621; homeo box C10; homeobox C10; homeobox protein Hox-3.6; Homeobox protein Hox-3I; homeobox protein Hox-C10; homeoprotein C10; Hox3.5 homeobox; Hox-3.6; HOX3I; HOXC10; Hoxc-10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 209448, 315338, 3226 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.57 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P31257, Q9NYD6 | |
| HOXC10 | |
| Recombinant protein encompassing a sequence within the center region of human Homeobox C10. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction