missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXC10 Monoclonal Antibody (OTI5C10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527065
This item is not returnable.
View return policy
Description
HOXC10 Monoclonal Antibody for Western Blot, IHC (P)
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The protein level is controlled during cell differentiation and proliferation, which may indicate this protein has a role in origin activation.
Specifications
| HOXC10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NYD6 | |
| HOXC10 | |
| Human recombinant protein fragment corresponding to amino acids 1-262 of HOXC10 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5C10 | |
| Unconjugated | |
| HOXC10 | |
| AI506621; homeo box C10; homeobox C10; homeobox protein Hox-3.6; Homeobox protein Hox-3I; homeobox protein Hox-C10; homeoprotein C10; Hox3.5 homeobox; Hox-3.6; HOX3I; HOXC10; Hoxc-10 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3226 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction