missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC10 Antibody (OTI5C10), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70909F
This item is not returnable.
View return policy
Description
HOXC10 Monoclonal specifically detects HOXC10 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| HOXC10 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| HOXC10 | |
| Recombinant protein fragment corresponding to amino acids 1-262 of human HOXC10 (NP_059105) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Immunohistochemistry | |
| OTI5C10 | |
| Immunohistochemistry | |
| homeo box C10, homeobox C10, Homeobox protein Hox-3I, homeobox protein Hox-C10, homeoprotein C10, HOX3I, MGC5259 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 3226.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction