missing translation for 'onlineSavingsMsg'
Learn More

HOXC10 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18409432 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409432 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409432 Supplier Novus Biologicals Supplier No. NBP171932AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HOXC10 Polyclonal antibody specifically detects HOXC10 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC10
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias homeo box C10, homeobox C10, Homeobox protein Hox-3I, homeobox protein Hox-C10, homeoprotein C10, HOX3I, MGC5259
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 50 and 100 of human Homeobox C10 using the numbering given in entry NP_059105.2 (GeneID 3226).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3226
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.