missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXB9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101087
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HOXB9.
HOXB9 belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. HOXB9 is one of several homeobox HOXB genes located in a cluster on chromosome 17. The exact role of this gene has yet to be determined.
Specifications
| HOXB9 | |
| Polyclonal | |
| Unconjugated | |
| HOXB9 | |
| HGNC:5120; homeo box 2E; homeo box B9; homeobox B9; homeobox protein Hox-2.5; Homeobox protein Hox-2E; homeobox protein Hox-B9; HOX2; HOX-2.5; HOX2E; HOXB9; Hoxb-9; MGC124051; MGC124052; MGC124053; MGC124054 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15417, 3219 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17482, P20615 | |
| HOXB9 | |
| A synthesized peptide derived from human HOXB9(Accession P17482), corresponding to amino acid residues S8-V58. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction