missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HOXB4 Recombinant Rabbit Monoclonal Antibody (K01_1S45)
GREENER_CHOICE

Product Code. 30827023
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827023 100 μL
30825524 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827023 Supplier Invitrogen™ Supplier No. MA563075

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

HOXB4 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

HOX homeobox genes play a role both in the early stem cell function as well as in later stages of hematopoietic differentiation. HOXB4, specifically, is a homeotic selector gene implicated in self-renewal of definitive HSCs. Expression of HOXB4 in primitive progenitors combined with culture on hematopoietic stroma induced a switch to the definitive HSC phenotype. Activation of WNT signaling in HSCs induced increased expression of HOXB4. Perturbations of HOX gene expression can cause leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXB4
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1S45
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene HOXB4
Gene Accession No. P17483
Gene Alias homeo box 2F; homeo box B4; homeo box B4a; homeobox B4; homeobox B4a; homeobox gene B-4; homeobox protein Hox-2.6; Homeobox protein Hox-2F; homeobox protein Hox-B4; Homeobox protein Hox-B4a; homeobox protein Zf-13; HOX2; Hox-2.6; HOX2F; Hoxb4; HOX-B4; Hoxb-4; Hoxb4 protein; hoxb4a; Hoxb4a variant 2; RGD1560113; sb:eu512; Z-17; zf13; ZF-13
Gene Symbols HOXB4
Host Species Rabbit
Immunogen A synthetic peptide of human HOXB4.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3214
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.