missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXB4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103140
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HoxB4.
HOX homeobox genes play a role both in the early stem cell function as well as in later stages of hematopoietic differentiation. HOXB4, specifically, is a homeotic selector gene implicated in self-renewal of definitive HSCs. Expression of HOXB4 in primitive progenitors combined with culture on hematopoietic stroma induced a switch to the definitive HSC phenotype. Activation of WNT signaling in HSCs induced increased expression of HOXB4. Perturbations of HOX gene expression can cause leukemia.
Specifications
| HOXB4 | |
| Polyclonal | |
| Unconjugated | |
| HOXB4 | |
| homeo box 2F; homeo box B4; homeo box B4a; homeobox B4; homeobox B4a; homeobox gene B-4; homeobox protein Hox-2.6; Homeobox protein Hox-2F; homeobox protein Hox-B4; Homeobox protein Hox-B4a; homeobox protein Zf-13; HOX2; Hox-2.6; HOX2F; Hoxb4; HOX-B4; Hoxb-4; Hoxb4 protein; hoxb4a; Hoxb4a variant 2; RGD1560113; sb:eu512; Z-17; zf13; ZF-13 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15412, 3214 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P10284, P17483 | |
| HOXB4 | |
| A synthesized peptide derived from human HOXB4(Accession P17483), corresponding to amino acid residues S4-F54. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction