missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103890
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HOXB3.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
Specifications
| HOXB3 | |
| Polyclonal | |
| Unconjugated | |
| HOXB3 | |
| homeo box 2G; homeo box B3; homeobox B3; homeobox protein Hox-2.7; Homeobox protein Hox-2G; homeobox protein Hox-B3; Homeobox protein MH-23; HOX2; Hox-2.7; HOX2G; HOXB3; Hoxb-3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15410, 303488, 3213 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09026, P14651 | |
| HOXB3 | |
| A synthesized peptide derived from human HOXB3(Accession P14651), corresponding to amino acid residues A359-D409. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction