missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539210
This item is not returnable.
View return policy
Description
Hoxb-1 maintains this function by acting very early during hindbrain neurogenesis to specify effectors of the sonic hedgehog and Mash1 signaling pathways. Hoxb2 is a homeodomain protein important in neural development that is also expressed during erythropoiesis, hindbrain development and normal human adult lung development. Hoxb2 may modulate the amount of gamma-globin mRNA expressed during development and differentiation. In addition, Hoxb2 plays an important role in the patterning of hindbrain and pharyngeal arches in the zebrafish.
Specifications
| HOXB2 | |
| Polyclonal | |
| Unconjugated | |
| HOXB2 | |
| Homeobox protein Hox-B2, Homeobox protein Hox-2.8, Homeobox protein Hox-2H, K8, HOX2H, HOXB2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3212 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P14652 | |
| HOXB2 | |
| A synthetic peptide derived from the internal region of human HOXB2 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction