missing translation for 'onlineSavingsMsg'
Learn More

HOXB2 Mouse anti-Human, Clone: PCRP-HOXB2-1F2, Novus Biologicals™

Product Code. 18381587 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381587 100 μg 100µg
18352822 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18381587 Supplier Novus Biologicals Supplier No. NBP313937100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HOXB2 Monoclonal antibody specifically detects HOXB2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXB2
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone PCRP-HOXB2-1F2
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P14652
Gene Alias homeo box 2H, homeo box B2, homeobox B2, Homeobox protein Hox-2.8, Homeobox protein Hox-2H, homeobox protein Hox-B2, HOX2, HOX2HHox-2.8, K8, K8 home protein
Host Species Mouse
Immunogen Recombinant fragment (aa114-220) of human HOXB2 (Uniprot: P14652)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3212
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.