missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB2 Antibody (PCRP-HOXB2-1F2), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14063MFV500
This item is not returnable.
View return policy
Description
HOXB2 Monoclonal antibody specifically detects HOXB2 in Human samples. It is validated for Flow Cytometry
Specifications
| HOXB2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 3212 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| PCRP-HOXB2-1F2 | |
| Flow Cytometry | |
| homeo box 2H, homeo box B2, homeobox B2, Homeobox protein Hox-2.8, Homeobox protein Hox-2H, homeobox protein Hox-B2, HOX2, HOX2HHox-2.8, K8, K8 home protein | |
| Recombinant fragment (aa114-220) of human HOXB2 (Uniprot: P14652) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction