missing translation for 'onlineSavingsMsg'
Learn More

HOXB2 Antibody (PCRP-HOXB2-1F2), HRP, Novus Biologicals™

Product Code. 18349352
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349352 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349352 Supplier Bio-Techne Supplier No. NBP314063H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HOXB2 Monoclonal antibody specifically detects HOXB2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXB2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-HOXB2-1F2
Conjugate HRP
Formulation PBS
Gene Alias homeo box 2H, homeo box B2, homeobox B2, Homeobox protein Hox-2.8, Homeobox protein Hox-2H, homeobox protein Hox-B2, HOX2, HOX2HHox-2.8, K8, K8 home protein
Host Species Mouse
Immunogen Recombinant fragment (aa114-220) of human HOXB2 (Uniprot: P14652)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3212
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.