missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXB13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117282
This item is not returnable.
View return policy
Description
Centrifuge briefly prior to opening.
HOXB13 encodes a transcription factor that belongs to the homeobox gene family. Genes of this family are highly conserved among vertebrates and essential for vertebrate embryonic development. This gene has been implicated to play a role in fetal skin development and cutaneous regeneration. In mice, a similar gene was shown to exhibit temporal and spatial colinearity in the main body axis of the embryo, but was not expressed in the secondary axes, which suggests functions in body patterning along the axis. This gene and other HOXB genes form a gene cluster at chromosome the 17q21-22 region.
Specifications
| HOXB13 | |
| Polyclonal | |
| Unconjugated | |
| HOXB13 | |
| homeo box B13; homeobox B13; homeobox protein Hox-B13; HOXA13; HOXB13; PSGD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10481 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q92826 | |
| HOXB13 | |
| Recombinant protein encompassing a sequence within the center region of human HOXB13. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction