missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXA9 Recombinant Rabbit Monoclonal Antibody (23GB6645), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | HOXA9 |
|---|---|
| Clone | 23GB6645 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088688
![]() |
Invitrogen™
MA553086 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089965
|
Invitrogen™
MA553087 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXA9 Recombinant Monoclonal antibody specifically detects HOXA9 in Human, Mouse samples. It is validated for Flow Cytometry, Western BlotSpecifications
| HOXA9 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P31269 | |
| 3205 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6645 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ABD-B; D6a9; homeo box A9; homeobox A9; homeobox A9-like; homeobox protein Hox-1.7; Homeobox protein Hox-1G; homeobox protein Hox-A9; homeodomain protein HOXA9; HOX1; HOX1.7; Hox-1.7; HOX1G; HOXA 9; Hoxa9; Hoxa-9; Hoxa-9 protein; Hoxa9l; RGD1310001 | |
| A synthesized peptide derived from human HOXA9 (231-272AA). | |
| Primary | |
| -20°C | |
| Hoxa9 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title